siRNA Details
Gene Symbol: Gm31169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK156678.1 | - | 5 | 1111 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY765785.1 | - | 3 | 209 | Chr. 6 - 143109214 - 143149134 [-] on Build GRCm38 |
