siRNA Details
Gene Symbol: Gm38958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131705.1 | - | 1 | 445 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706152.1 | - | 1 | 438 | Chr. 11 - 95823957 - 95824405 [-] on Build GRCm38 |
