siRNA Details
Gene Symbol: 2900003A17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013479.1 | - | 1 | 437 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712445.1 | - | 1 | 437 | Chr. 12 - 25061784 - 25062314 [+] on Build GRCm38 |
