siRNA Details
Gene Symbol: 4930402F11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045940.1 | 3 | 372 | |||
GenBank mRNA | AK015048.1 | - | 3 | 374 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714574.1 | - | 374 | Chr. - - [-] on Build |
