siRNA Details
Gene Symbol: Cfap54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015623.1 | - | 1 | 245 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715174.1 | - | 1 | 246 | Chr. 10 - 93081306 - 93081902 [+] on Build GRCm38 |
