siRNA Details
Gene Symbol: Gm21284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_078345.1 | 8 | 1345 | |||
GenBank mRNA | AK137133.1 | - | 8 | 1346 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm21284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_078345.1 | 8 | 1345 | |||
GenBank mRNA | AK137133.1 | - | 8 | 1346 |