siRNA Details
Gene Symbol: Gm21284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_078345.1 | 2 | 166 | |||
GenBank mRNA | AK137133.1 | - | 2 | 167 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB627504.1 | - | 2 | 167 | Chr. 6 - 83568039 - 83580106 [+] on Build GRCm38 |
