siRNA Details
Gene Symbol: C130057N11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK048403.1 | - | 1 | 3404 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB371202.2 | - | 1 | 330 | Chr. 2 - 154870297 - 154871010 [+] on Build GRCm38 |
