siRNA Details
Gene Symbol: Zim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007312.2 | - | 532 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707418.1 | - | 3 | 532 | Chr. 7 - 6976302 - 6977420 [-] on Build GRCm38 |
