siRNA Details
Gene Symbol: E130102B10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021366.1 | - | 1 | 288 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY731236.1 | - | 1 | 288 | Chr. 16 - 75010687 - 75011090 [-] on Build GRCm38 |
