siRNA Details
Gene Symbol: 4930568E02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016249.1 | - | 1 | 472 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715861.1 | - | 1 | 468 | Chr. 17 - 25183812 - 25184456 [-] on Build GRCm38 |
