siRNA Details
Gene Symbol: Gm11730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI154020.1 | - | 3 | 320 | ||
AK086245.1 | - | 2 | 550 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB523703.2 | - | 433 | Chr. - - [-] on Build |
