siRNA Details
Gene Symbol: 5033424D13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017190.1 | - | 1 | 339 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716920.1 | - | 1 | 335 | Chr. 12 - 108878772 - 108879418 [+] on Build GRCm38 |
