siRNA Details
Gene Symbol: 4930543I03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016029.1 | - | 3 | 225 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715609.1 | - | 3 | 225 | Chr. 15 - 86737826 - 86766127 [+] on Build GRCm38 |
