siRNA Details
Gene Symbol: Gm16617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045728.1 | 3 | 455 | |||
GenBank mRNA | AK136665.1 | - | 3 | 456 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721044.1 | - | 3 | 453 | Chr. 14 - 51986425 - 51988827 [-] on Build GRCm38 |
