siRNA Details
Gene Symbol: 4930542M03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016022.1 | - | 1 | 77 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715601.1 | - | 1 | 74 | Chr. 17 - 45879849 - 45881440 [-] on Build GRCm38 |
