siRNA Details
Gene Symbol: 9430083B18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020503.1 | - | 1 | 469 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721880.1 | - | 1 | 469 | Chr. 1 - 172873551 - 172874203 [+] on Build GRCm38 |
