siRNA Details
Gene Symbol: 2510017J16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK010976.1 | - | 1 | 643 | ||
AK010977.1 | - | 1 | 643 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY710361.1 | - | 1 | 643 | Chr. 5 - 3585112 - 3586053 [+] on Build GRCm38 |
