siRNA Details
Gene Symbol: Gm13974 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043709.1 | - | 9 | 1268 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB265936.2 | - | 4 | 480 | Chr. 2 - 114975470 - 115043961 [+] on Build GRCm38 |
