siRNA Details
Gene Symbol: Gm4117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087351.1 | - | 545 | |||
BB536129.2 | - | 477 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB664604.1 | - | 2 | 545 | Chr. 13 - 89739464 - 89887078 [+] on Build GRCm38 |
