siRNA Details
Gene Symbol: 1700021F02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006198.1 | - | 2 | 218 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706294.1 | - | 2 | 211 | Chr. 5 - 103346601 - 103348299 [+] on Build GRCm38 |
