siRNA Details
Gene Symbol: 1700084K02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006995.1 | - | 795 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV206122.1 | - | 1 | 212 | Chr. 19 - 43940621 - 43940920 [+] on Build GRCm38 |
