siRNA Details
Gene Symbol: 1110055C04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004262.1 | - | 1 | 305 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY704505.1 | - | 1 | 301 | Chr. 11 - 60796310 - 60797241 [+] on Build GRCm38 |
