siRNA Details
Gene Symbol: Gm33172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142521.1 | - | 3 | 625 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB660869.1 | - | 3 | 625 | Chr. 13 - 110683912 - 110686229 [+] on Build GRCm38 |
