siRNA Details
Gene Symbol: 4930527A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015909.1 | - | 1 | 298 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715466.1 | - | 1 | 295 | Chr. 2 - 105476504 - 105477357 [+] on Build GRCm38 |
