siRNA Details
Gene Symbol: 4933425B07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046460.1 | 5 | 628 | |||
GenBank mRNA | AK016907.1 | - | 5 | 629 | ||
BY716586.1 | - | 5 | 629 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933425B07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046460.1 | 5 | 628 | |||
GenBank mRNA | AK016907.1 | - | 5 | 629 | ||
BY716586.1 | - | 5 | 629 |