siRNA Details
Gene Symbol: 1600029O15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033522.1 | 3 | 1708 | |||
GenBank mRNA | AK005564.2 | - | 3 | 1707 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1600029O15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033522.1 | 3 | 1708 | |||
GenBank mRNA | AK005564.2 | - | 3 | 1707 |