siRNA Details
Gene Symbol: Gm34888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034386.1 | - | 3 | 399 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB626421.1 | - | 3 | 399 | Chr. 9 - 109537100 - 109550818 [+] on Build GRCm38 |
