siRNA Details
Gene Symbol: Gm29128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139037.1 | - | 2 | 220 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY724809.1 | - | 2 | 220 | Chr. 1 - 31117139 - 31136638 [+] on Build GRCm38 |
