siRNA Details
Gene Symbol: Sh3glb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052668.1 | - | 1 | 1269 | ||
AK086477.1 | - | 1 | 1209 | |||
AK147816.1 | - | 1 | 1246 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB194902.1 | - | 1 | 42 | Chr. 3 - 144687235 - 144688066 [-] on Build GRCm38 |
