siRNA Details
Gene Symbol: 4933411E08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045342.1 | 3 | 309 | |||
GenBank mRNA | AK016769.1 | - | 3 | 310 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716447.1 | - | 3 | 310 | Chr. 10 - 117925459 - 117947240 [+] on Build GRCm38 |
