siRNA Details
Gene Symbol: Gm12069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK158601.1 | - | 2 | 155 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY240988.1 | - | 2 | 152 | Chr. 11 - 25301263 - 25322448 [+] on Build GRCm38 |
