siRNA Details
Gene Symbol: Gm33255 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135250.1 | - | 3 | 400 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ051271.1 | - | 3 | 400 | Chr. 16 - 88368238 - 88381696 [-] on Build GRCm38 |
