siRNA Details
Gene Symbol: Gm31503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK145338.1 | - | 345 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY735551.1 | - | 2 | 345 | Chr. 18 - 83363372 - 83371350 [-] on Build GRCm38 |
