siRNA Details
Gene Symbol: Zic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141431.1 | - | 1 | 1822 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK780542.1 | - | 1 | 56 | Chr. 9 - 91358476 - 91359230 [-] on Build GRCm38 |
