siRNA Details
Gene Symbol: 6230424C14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020242.1 | - | 1 | 26 | ||
AK078961.1 | - | 1 | 1309 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CX224993.1 | - | 1 | 709 | Chr. 13 - 114953388 - 114954177 [-] on Build GRCm38 |
