siRNA Details
Gene Symbol: 1600020E01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037960.1 | 3, 4 | 312 | |||
NR_037961.1 | 3, 4 | 339 | ||||
GenBank mRNA | AA162444.1 | - | 3, 4 | 310 | ||
AK005503.1 | - | 3, 4 | 340 | |||
BC029726.1 | - | 1829 |
