siRNA Details
Gene Symbol: 4930579K21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076967.1 | - | 1 | 102 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB615956.1 | - | 1 | 100 | Chr. 11 - 50723774 - 50725542 [-] on Build GRCm38 |
