siRNA Details
Gene Symbol: A930016I07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020866.1 | - | 1 | 836 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB283723.1 | - | 1 | 491 | Chr. 7 - 114364336 - 114364872 [-] on Build GRCm38 |
