siRNA Details
Gene Symbol: 1700012D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047116.1 | - | 1 | 1516 | ||
AK087602.1 | - | 1 | 1505 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700012D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047116.1 | - | 1 | 1516 | ||
AK087602.1 | - | 1 | 1505 |