siRNA Details
Gene Symbol: 4922502B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015021.1 | - | 1217 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB679327.1 | - | 1 | 413 | Chr. 8 - 109250481 - 109250925 [-] on Build GRCm38 |
