siRNA Details
Gene Symbol: 4922502B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015021.1 | - | 478 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714560.1 | - | 1 | 475 | Chr. 8 - 109250850 - 109251720 [-] on Build GRCm38 |
