siRNA Details
Gene Symbol: Hif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK048798.1 | - | 434 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB650202.1 | - | 2 | 434 | Chr. 12 - 73933509 - 73949785 [-] on Build GRCm38 |
