siRNA Details
Gene Symbol: Kazn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034042.1 | - | 1 | 1955 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB078061.2 | - | 1 | 166 | Chr. 4 - 142137924 - 142138586 [-] on Build GRCm38 |
