siRNA Details
Gene Symbol: A730082K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040317.1 | 4 | 1408 | |||
NR_040319.1 | 5 | 1386 | ||||
GenBank mRNA | AK043303.1 | - | 2 | 956 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A730082K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040317.1 | 4 | 1408 | |||
NR_040319.1 | 5 | 1386 | ||||
GenBank mRNA | AK043303.1 | - | 2 | 956 |