siRNA Details
Gene Symbol: A730082K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040317.1 | 4 | 1214 | |||
NR_040319.1 | 5 | 1192 | ||||
GenBank mRNA | AK043303.1 | - | 2 | 762 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DV661602.1 | - | 1 | 175 | Chr. 7 - 114983192 - 114983960 [+] on Build GRCm38 |
