siRNA Details
Gene Symbol: A730082K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040317.1 | 4 | 617 | |||
NR_040319.1 | 5 | 595 | ||||
GenBank mRNA | AK043303.1 | - | 2 | 165 | ||
BG082928.2 | - | 5 | 562 | |||
DV654286.1 | - | 4 | 617 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DV663227.1 | - | 4 | 616 | Chr. 7 - 114948651 - 114982921 [+] on Build GRCm38 |
