siRNA Details
Gene Symbol: Grip1os2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045359.1 | 3 | 394 | |||
GenBank mRNA | AK016220.1 | - | 3 | 394 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715827.1 | - | 394 | Chr. - - [-] on Build |
