siRNA Details
Gene Symbol: 1700026H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK138516.1 | - | 1 | 650 | ||
AK138667.1 | - | 1 | 650 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY724251.1 | - | 1 | 650 | Chr. 10 - 20170780 - 20171467 [+] on Build GRCm38 |
