siRNA Details
Gene Symbol: 9430095M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020525.1 | - | 1 | 238 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721949.1 | - | 1 | 235 | Chr. 4 - 55020913 - 55021286 [+] on Build GRCm38 |
